Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
TXNDC9
Entrez Gene ID
101106316
Gene Name
thioredoxin domain containing 9
Gene Aliases
-
Chromosome Location
Chr.X: 98770122 - 98782066 on Build Oar_rambouillet_v1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_004006137.4 XP_004006186.2 2 - 3 356 64

View Full Product Details