Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
CNFN
Entrez Gene ID
101104084
Gene Name
cornifelin
Gene Aliases
-
Chromosome Location
Chr.14: 54079859 - 54082175 on Build Oar_rambouillet_v1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_012190469.2 XP_012045859.1 2 - 3 200 96
XM_027978398.1 XP_027834199.1 3 - 4 330 96
XM_027978399.1 XP_027834200.1 3 - 4 353 96
XM_027978400.1 XP_027834201.1 3 - 4 370 96

View Full Product Details