Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
POLR1G
Entrez Gene ID
101113091
Gene Name
RNA polymerase I subunit G
Gene Aliases
CD3EAP
Chromosome Location
Chr.14: 56513386 - 56516140 on Build Oar_rambouillet_v1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_004015318.4 XP_004015367.2 2 - 3 365 85

View Full Product Details