Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
PMCH
Entrez Gene ID
443522
Gene Name
pro-melanin concentrating hormone
Gene Aliases
-
Chromosome Location
Chr.3: 183707644 - 183709286 on Build Oar_rambouillet_v1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_004006673.4 XP_004006722.1 1 - 2 746 119
GenBank mRNA AY603756.1 - 1 - 2 166 119

View Full Product Details