Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
PHF5A
Entrez Gene ID
101110927
Gene Name
PHD finger protein 5A
Gene Aliases
-
Chromosome Location
Chr.3: 233419917 - 233428878 on Build Oar_rambouillet_v1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_004007013.3 XP_004007062.1 2 - 3 118 66

View Full Product Details