Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
MEOX2
Entrez Gene ID
101103559
Gene Name
mesenchyme homeobox 2
Gene Aliases
-
Chromosome Location
Chr.4: 27298374 - 27374399 on Build Oar_rambouillet_v1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_004007766.4 XP_004007815.2 2 - 3 995 80

View Full Product Details