Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
GRM3
Entrez Gene ID
101113057
Gene Name
glutamate metabotropic receptor 3
Gene Aliases
-
Chromosome Location
Chr.4: 37916900 - 38160326 on Build Oar_rambouillet_v1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_004007802.4 XP_004007851.2 3 - 4 1746 109

View Full Product Details