Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
PPCS
Entrez Gene ID
100145879
Gene Name
phosphopantothenoylcysteine synthetase
Gene Aliases
DFP
Chromosome Location
Chr.1: 17605579 - 17609062 on Build Oar_rambouillet_v1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001126363.1 NP_001119835.1 1 - 2 505 55
XM_027970081.1 XP_027825882.1 1 - 2 550 55
GenBank mRNA EU583471.1 - 1 - 2 505 55

View Full Product Details