Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
UTP4
Entrez Gene ID
101120686
Gene Name
UTP4 small subunit processome component
Gene Aliases
CIRH1A
Chromosome Location
Chr.14: 39117136 - 39146973 on Build Oar_rambouillet_v1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_004015097.4 XP_004015146.2 9 - 10 1187 99
XM_012190032.3 XP_012045422.2 9 - 10 1126 99

View Full Product Details