The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | XM_027973525.1 | XP_027829326.1 | 4 - 5 | 693 | 59 |
GenBank mRNA | EF121765.1 | - | 3 - 4 | 311 | 59 |