Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
VWC2L
Entrez Gene ID
101104637
Gene Name
von Willebrand factor C domain containing 2 like
Gene Aliases
-
Chromosome Location
Chr.2: 231108622 - 231297704 on Build Oar_rambouillet_v1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_027965154.1 XP_027820955.1 1 - 2 390 106

View Full Product Details