Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
TYR
Entrez Gene ID
780502
Gene Name
tyrosinase
Gene Aliases
-
Chromosome Location
Chr.21: 7263508 - 7379924 on Build Oar_rambouillet_v1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001130027.1 NP_001123499.1 4 - 5 1393 146
GenBank mRNA EU760772.1 - 4 - 5 1393 146

View Full Product Details