Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
IL7
Entrez Gene ID
443341
Gene Name
interleukin 7
Gene Aliases
-
Chromosome Location
Chr.9: 59923896 - 59981524 on Build Oar_rambouillet_v1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001009777.1 NP_001009777.1 5 - 6 496 78
GenBank mRNA U10089.1 - 5 - 6 496 78

View Full Product Details