Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
PLAUR
Entrez Gene ID
100302564
Gene Name
plasminogen activator, urokinase receptor
Gene Aliases
u-PAR
Chromosome Location
Chr.14: 55382650 - 55395939 on Build Oar_rambouillet_v1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001163606.1 NP_001157078.1 5 - 6 641 83
XM_012189516.3 XP_012044906.2 5 - 6 662 83
GenBank mRNA FJ803200.1 - 5 - 6 641 83

View Full Product Details