Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
TLR3
Entrez Gene ID
554258
Gene Name
toll like receptor 3
Gene Aliases
-
Chromosome Location
Chr.26: 17145731 - 17156798 on Build Oar_rambouillet_v1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001135928.1 NP_001129400.1 3 - 4 712 117
XM_012105647.3 XP_011961037.2 3 - 4 1036 117
GenBank mRNA AM981301.1 - 3 - 4 712 117

View Full Product Details