Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
GATD3A
Entrez Gene ID
100302318
Gene Name
glutamine amidotransferase like class 1 domain containing 3A
Gene Aliases
C1H21orf33, C21orf33
Chromosome Location
Chr.1: 287477779 - 287485863 on Build Oar_rambouillet_v1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001162560.1 NP_001156032.1 5 - 6 541 101
GenBank mRNA FJ969404.1 - 5 - 6 541 101

View Full Product Details