Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
GATA1
Entrez Gene ID
100820747
Gene Name
GATA binding protein 1
Gene Aliases
-
Chromosome Location
Chr.X: 58599341 - 58602122 on Build Oar_rambouillet_v1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001252179.1 NP_001239108.1 2 - 3 597 71
GenBank mRNA AB612140.1 - 2 - 3 597 71

View Full Product Details