Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
SQSTM1
Entrez Gene ID
100135450
Gene Name
sequestosome 1
Gene Aliases
-
Chromosome Location
Chr.5: 1347420 - 1357549 on Build Oar_rambouillet_v1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001163043.1 NP_001156515.1 4 - 5 677 94
GenBank mRNA FJ958359.1 - 4 - 5 677 94

View Full Product Details