Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
SPX
Entrez Gene ID
101118938
Gene Name
spexin hormone
Gene Aliases
C3H12orf39
Chromosome Location
Chr.3: 207828921 - 207838391 on Build Oar_rambouillet_v1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001287469.1 NP_001274398.1 4 - 5 247 90
XM_027965740.1 XP_027821541.1 3 - 4 472 90
GenBank mRNA KF408269.1 - 4 - 5 247 90

View Full Product Details