Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
CD93
Entrez Gene ID
100302099
Gene Name
CD93 molecule
Gene Aliases
-
Chromosome Location
Chr.13: 43298859 - 43301040 on Build Oar_rambouillet_v1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001161888.1 NP_001155360.1 1 - 1 920 106
GenBank mRNA FJ958365.1 - 1 - 1 920 106

View Full Product Details