Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
SH2D1B
Entrez Gene ID
101108790
Gene Name
SH2 domain containing 1B
Gene Aliases
-
Chromosome Location
Chr.1: 121065616 - 121095940 on Build Oar_rambouillet_v1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_004002709.4 XP_004002758.1 1 - 2 252 95
XM_012185125.3 XP_012040515.1 1 - 2 252 95

View Full Product Details