Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
PIN1
Entrez Gene ID
101110756
Gene Name
peptidylprolyl cis/trans isomerase, NIMA-interacting 1
Gene Aliases
-
Chromosome Location
Chr.5: 12706392 - 12718526 on Build Oar_rambouillet_v1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_004008507.4 XP_004008556.2 2 - 3 324 99

View Full Product Details