Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
MANEA
Entrez Gene ID
101106778
Gene Name
mannosidase endo-alpha
Gene Aliases
-
Chromosome Location
Chr.10: 501005 - 582241 on Build Oar_rambouillet_v1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_004011265.4 XP_004011314.2 3 - 4 790 142

View Full Product Details