Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
RGS17
Entrez Gene ID
101104247
Gene Name
regulator of G protein signaling 17
Gene Aliases
-
Chromosome Location
Chr.8: 84205410 - 84240374 on Build Oar_rambouillet_v1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_015097491.2 XP_014952977.2 2 - 3 225 79

View Full Product Details