The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | XM_004015097.4 | XP_004015146.2 | 6 - 7 | 825 | 59 |
XM_012190032.3 | XP_012045422.2 | 6 - 7 | 764 | 59 |