Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
ZBTB47
Entrez Gene ID
101119338
Gene Name
zinc finger and BTB domain containing 47
Gene Aliases
-
Chromosome Location
Chr.19: 15710488 - 15722729 on Build Oar_rambouillet_v1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_027958061.1 XP_027813862.1 4 - 5 1948 93

View Full Product Details