Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
NEFL
Entrez Gene ID
101119539
Gene Name
neurofilament light
Gene Aliases
-
Chromosome Location
Chr.2: 42835923 - 42841650 on Build Oar_rambouillet_v1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_015093090.2 XP_014948576.2 2 - 3 1348 96

View Full Product Details