Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
CSRNP1
Entrez Gene ID
101112518
Gene Name
cysteine and serine rich nuclear protein 1
Gene Aliases
-
Chromosome Location
Chr.19: 13060315 - 13072727 on Build Oar_rambouillet_v1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_027958026.1 XP_027813827.1 3 - 4 6754 88
XM_027958027.1 XP_027813828.1 4 - 5 977 88

View Full Product Details