Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
HAUS3
Entrez Gene ID
101110940
Gene Name
HAUS augmin like complex subunit 3
Gene Aliases
-
Chromosome Location
Chr.6: 128442912 - 128456644 on Build Oar_rambouillet_v1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_004023443.4 XP_004023492.2 2 - 3 1468 101
XM_027971301.1 XP_027827102.1 3 - 4 1139 101

View Full Product Details