Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
LYVE1
Entrez Gene ID
101103009
Gene Name
lymphatic vessel endothelial hyaluronan receptor 1
Gene Aliases
-
Chromosome Location
Chr.15: 45145489 - 45160457 on Build Oar_rambouillet_v1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_015101033.2 XP_014956519.2 2 - 3 454 73

View Full Product Details