Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
ZFP90
Entrez Gene ID
101111386
Gene Name
ZFP90 zinc finger protein
Gene Aliases
-
Chromosome Location
Chr.14: 38659424 - 38696788 on Build Oar_rambouillet_v1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_012190017.2 XP_012045407.2 2 - 3 271 71
XM_027978116.1 XP_027833917.1 3 - 4 241 71
XM_027978117.1 XP_027833918.1 3 - 4 261 71

View Full Product Details