Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
PANX3
Entrez Gene ID
101106978
Gene Name
pannexin 3
Gene Aliases
-
Chromosome Location
Chr.21: 29088706 - 29097138 on Build Oar_rambouillet_v1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_004019501.4 XP_004019550.1 3 - 4 627 79

View Full Product Details