Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
IL36A
Entrez Gene ID
101106251
Gene Name
interleukin 36 alpha
Gene Aliases
-
Chromosome Location
Chr.3: 63999275 - 64003864 on Build Oar_rambouillet_v1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_012170041.3 XP_012025431.1 3 - 4 506 85
XM_012170048.3 XP_012025438.1 3 - 4 500 85
XM_027966771.1 XP_027822572.1 2 - 3 325 85

View Full Product Details