Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
DLX3
Entrez Gene ID
101109372
Gene Name
distal-less homeobox 3
Gene Aliases
-
Chromosome Location
Chr.11: 26663035 - 26668163 on Build Oar_rambouillet_v1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_004012779.4 XP_004012828.2 1 - 2 502 97

View Full Product Details