Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
BEND7
Entrez Gene ID
101115641
Gene Name
BEN domain containing 7
Gene Aliases
-
Chromosome Location
Chr.13: 29123072 - 29207809 on Build Oar_rambouillet_v1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_027976641.1 XP_027832442.1 3 - 4 2725 78
XM_027976642.1 XP_027832443.1 3 - 4 2726 78
XM_027976643.1 XP_027832444.1 4 - 5 599 78
XM_027976644.1 XP_027832445.1 4 - 5 599 78
XM_027976645.1 XP_027832446.1 3 - 4 2724 78
XM_027976647.1 XP_027832448.1 3 - 4 2723 78
XM_027976648.1 XP_027832449.1 3 - 4 2724 78
XM_027976649.1 XP_027832450.1 3 - 4 496 78

View Full Product Details