Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
HTR2B
Entrez Gene ID
101106827
Gene Name
5-hydroxytryptamine receptor 2B
Gene Aliases
-
Chromosome Location
Chr.2: 248514839 - 248530877 on Build Oar_rambouillet_v1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_012148490.3 XP_012003880.1 3 - 4 1739 108
XM_012148499.3 XP_012003889.1 3 - 4 1378 108

View Full Product Details