Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
PPDPF
Entrez Gene ID
101109634
Gene Name
pancreatic progenitor cell differentiation and proliferation factor
Gene Aliases
-
Chromosome Location
Chr.13: 56437455 - 56439087 on Build Oar_rambouillet_v1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_027976944.1 XP_027832745.1 4 - 5 708 83

View Full Product Details