Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
GLTP
Entrez Gene ID
101113276
Gene Name
glycolipid transfer protein
Gene Aliases
-
Chromosome Location
Chr.17: 72579755 - 72600560 on Build Oar_rambouillet_v1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_027956635.1 XP_027812436.1 3 - 4 449 78

View Full Product Details