Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
ALKBH1
Entrez Gene ID
101111374
Gene Name
alkB homolog 1, histone H2A dioxygenase
Gene Aliases
-
Chromosome Location
Chr.7: 92809718 - 92835324 on Build Oar_rambouillet_v1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_004011087.3 XP_004011136.2 4 - 5 592 79
XM_027972084.1 XP_027827885.1 4 - 5 562 79
XM_027972085.1 XP_027827886.1 4 - 5 601 79

View Full Product Details