Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
LCA5L
Entrez Gene ID
101123084
Gene Name
lebercilin LCA5 like
Gene Aliases
-
Chromosome Location
Chr.1: 282581118 - 282607925 on Build Oar_rambouillet_v1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_012103821.3 XP_011959211.2 6 - 7 1436 92

View Full Product Details