Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
ELOVL3
Entrez Gene ID
101106896
Gene Name
ELOVL fatty acid elongase 3
Gene Aliases
-
Chromosome Location
Chr.22: 25493316 - 25496456 on Build Oar_rambouillet_v1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_012102841.3 XP_011958231.1 1 - 2 286 66

View Full Product Details