Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
DPEP2
Entrez Gene ID
101110863
Gene Name
dipeptidase 2
Gene Aliases
-
Chromosome Location
Chr.14: 38232413 - 38239840 on Build Oar_rambouillet_v1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_027978104.1 XP_027833905.1 11 - 12 1408 72

View Full Product Details