Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
PATE2
Entrez Gene ID
101117712
Gene Name
prostate and testis expressed 2
Gene Aliases
-
Chromosome Location
Chr.21: 30206495 - 30207778 on Build Oar_rambouillet_v1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_004019824.4 XP_004019873.3 3 - 4 201 99

View Full Product Details