Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
TIGIT
Entrez Gene ID
101116533
Gene Name
T cell immunoreceptor with Ig and ITIM domains
Gene Aliases
-
Chromosome Location
Chr.1: 196578723 - 196596304 on Build Oar_rambouillet_v1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_027962473.1 XP_027818274.1 2 - 3 599 56

View Full Product Details