Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
COG8
Entrez Gene ID
101113170
Gene Name
component of oligomeric golgi complex 8
Gene Aliases
-
Chromosome Location
Chr.14: 39248160 - 39254810 on Build Oar_rambouillet_v1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_027978123.1 XP_027833924.1 3 - 4 1454 86

View Full Product Details