Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
PROCR
Entrez Gene ID
101115814
Gene Name
protein C receptor
Gene Aliases
-
Chromosome Location
Chr.13: 67245312 - 67250112 on Build Oar_rambouillet_v1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_012189121.3 XP_012044511.2 1 - 2 239 62
XM_015099846.2 XP_014955332.2 2 - 3 231 62

View Full Product Details