Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
KRT5
Entrez Gene ID
100303605
Gene Name
keratin 5
Gene Aliases
-
Chromosome Location
Chr.3: 143379949 - 143385944 on Build Oar_rambouillet_v1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_027967278.1 XP_027823079.1 4 - 5 933 88
GenBank mRNA GQ372827.1 - 3 - 4 310 88

View Full Product Details