Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
NUDT19
Entrez Gene ID
101109295
Gene Name
nudix hydrolase 19
Gene Aliases
-
Chromosome Location
Chr.14: 46084516 - 46091658 on Build Oar_rambouillet_v1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_004015139.4 XP_004015188.1 1 - 2 875 81

View Full Product Details