Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
PLAC9
Entrez Gene ID
101106391
Gene Name
placenta associated 9
Gene Aliases
-
Chromosome Location
Chr.25: 36370845 - 36382385 on Build Oar_rambouillet_v1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_004021513.4 XP_004021562.2 1 - 2 1584 75

View Full Product Details