Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
FAM20A
Entrez Gene ID
101108764
Gene Name
FAM20A golgi associated secretory pathway pseudokinase
Gene Aliases
-
Chromosome Location
Chr.11: 1652228 - 1695763 on Build Oar_rambouillet_v1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_027973995.1 XP_027829796.1 8 - 9 1232 69
XM_027973996.1 XP_027829797.1 6 - 7 1903 69
XM_027973997.1 XP_027829798.1 8 - 9 956 69

View Full Product Details